missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Human MAPK14 (p38 alpha), GST Tag Recombinant Protein
Human MAPK14 (p38 alpha), GST Tag Recombinant Protein
Supplier: Thermo Fisher Scientific PV3304
Description
MAPK14 (p38 alpha) is a member of the stress-activated protein kinase class of MAPKs.Specifications
NP_620581 | |
1432 | |
10μg | |
MAP Kinase Family | |
Recombinant | |
Storage Buffer: 20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CMGC Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
mitogen-activated protein kinase 14 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
67.6 kDa | |
MAPK14 (p38 α) | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
MAPK14 | |
E. coli | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
For Research Use Only. Not for use in diagnostic procedures.