MAPA™ Ultrane™ 559 Foam Nitrile Coated Gloves
Feature an ASTM Cut Level 1 HDPE liner
$12.25 - $13.80
Specifications
Cut Resistance Rating Level 1
Material Nitrile foam
View More Specs
Products 4
Catalog Number Mfr. No. Color Size Price Quantity  
Catalog Number Mfr. No. Color Size Price Quantity  
19-249-733 View Documents MAPA™
559419
Gray 9 Pair for $12.25
Case of 48 Pair for $548.64
19-249-732 View Documents MAPA™
559418
Gray 8 Pair for $12.25
Case of 48 Pair for $548.64
19-249-730 View Documents MAPA™
559410
Gray 10 Pair for $12.25
Case of 48 Pair for $548.64
19-249-805 View Documents MAPA™
559411
Black 11 N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
For handling glass window panes, sheet metal applications, and any operation with a moderate risk of cuts and contact with oily/wet parts.
  • Combined effect of dexterity and protection against mechanical hazards
  • Fully coated foam nitrile coating with HDPE and polyamide liner
  • Excellent abrasion resistance along with exceptional comfort
  • Smooth finish with fully coated back and fingers
  • Hand-specific design in four sizes for optimum fit
  • Can be reused after washing at 40°C (104°F) with standard detergent
Compliance
ANSI™ Level 1 cut protection
Specifications
Specifications
Level 1
Nitrile foam