Please login to your online account to display your discounted pricing
MAPA™ TitanLite™ Gloves
Outlasts and outperforms traditional work gloves
$3.08 - $3.08
Products
Catalog Number Mfr. No. Price Quantity    
28-709-38
mapa™
397427
Pair for $3.08
Case of 100 Pair for $276.00
Description & Specifications
For light- to medium-duty applications.
  • Comfortable interlock cotton liner has a light coating of nitrile for superior resistance to cuts, snags, punctures, and abrasion
  • Coated on the palm and fingers only, leaving the back of the glove uncoated for excellent ventilation and cool hands
  • Anatomically formed to the shape of the hand, reducing hand fatigue
  • Treated with Actifresh™ for resistance to bacteria, molds, and mildew. Launderable, with a smooth grip
  • With knitwrist
Meet CE standards.