Please login to your online account to display your discounted pricing

MAPA™ Stanflex™ Vinyl Bisque Grip Gloves

Combining chemical/mechanical protection with dexterity

$3.59 - $3.59

Specifications

Certifications CE
Color Yellow
Length (English) 10.5 in.
Length (Metric) 26.7cm
Material Vinyl
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

11-388-117D

 
mapa™
543429
X-Large Pair for $3.59
Case of 72 Pair for $235.44
Description & Specifications

Specifications

Certifications CE
Color Yellow
Length (English) 10.5 in.
Length (Metric) 26.7cm
Material Vinyl
Exterior Finish Bisque
For Use With (Application) intricate assembly, film processing, biological research, Resin application, Epoxy lay-up, furniture finishing
Glove Cuff Style Pinked
Hand Hand-specific
Powdered Powdered
Thickness (English) 20 mil

Designed for activities such as intricate assembly, film processing, biological research, resin application, epoxy layup, and furniture finishing. Liquid-tight gloves outperform natural rubber and nitrile in acids, caustics, alcohols, aldehydes, oils and grease.

  • Nonmigrating plasticizer prevents plasticizer blush, enhancing long life and flexibility while minimizing odor
  • Lightly powdered for easy donning and doffing
  • Hand-specific with curved finger design for low hand fatigue
  • Feature bisque grip and pinked cuffs
  • Thickness: 20mil

CE approved.