missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MAPA™ Solo Ultra 997™ Powder-Free Disposable Nitrile Gloves

Superior wet or dry grip

Manufacturer:  MAPA™ 997436

 View more versions of this product

Catalog No. 19-050-301B




Description

Description

Special nitrile formulation offers resistance to punctures and snags while protecting against a wide range of chemicals.

  • Made of 100% nitrile, with no natural rubber latex proteins that can cause allergic reactions
  • Powder-free, reducing the risk of dermatitis
  • Textured fingertips provide a sure wet or dry grip
  • Rolled cuff minimizes the chance of tearing while donning
  • Length 10 in. (25cm)
  • Gauge: 4mil

Order Info

Gloves packaged in dispenser box.

Compliance

Meet 21CFR170-199 CE approved.

Specifications

Specifications

Yes
Rolled
Small
Blue
24 cm
4 mil
Nitrile
Powder-free
9.5 in.
21 CFR 170-199, CE
SDS
Documents