MAPA™ Pylox™ Vinyl Bisque Grip Gloves
Combining chemical and mechanical protection with dexterity
$112.25 - $1,098.74
Specifications
Certifications CE
Color Yellow
Length (English) 10.5 in.
Length (Metric) 26.7cm
Material Vinyl
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Hand Price Quantity  
Catalog Number Mfr. No. Hand Price Quantity  
11-393-90B MAPA™
541428
Left Pack of 100 Each for $112.25
11-393-91B MAPA™
541978
Right Pack of 100 Each for $112.25
11-393-90 MAPA™
541426
Left Case of 1000 Each for $1,098.74
11-393-90A MAPA™
541427
Left Case of 1000 Each for $1,098.74
11-393-91 MAPA™
541976
Right Case of 1000 Each for $1,098.74
11-393-91A MAPA™
541977
Right Case of 1000 Each for $1,098.74
Specifications
Description
Designed for activities such as intricate assembly, film processing, biological research, resin application, epoxy layup and furniture finishing. Liquid-tight gloves outperform natural rubber and nitrile in acids, caustics, alcohols, aldehydes, oils and grease.
  • Nonmigrating plasticizer prevents plasticizer blush, enhancing long life and flexibility while minimizing odor
  • Lightly powdered for easy donning and doffing
  • Hand-specific with curved finger design for low hand fatigue
  • Feature bisque grip and pinked cuffs
  • Thickness: 5mil
Certifications
CE approved.
Description & Specifications
Specifications
CE
10.5 in.
Vinyl
Bisque
Pinked
5 mil
Yellow
26.7cm
Small
intricate assembly, film processing, biological research, Resin application, Epoxy layup and furniture finishing, liquid tight gloves outperform natural Rubber and Nitrile in Acids, Caustics, Alcohols, Aldehydes, Oils and G
Powdered