Please login to your online account to display your discounted pricing
MAPA™ Pylox™ Vinyl Bisque Grip Gloves
Combining chemical and mechanical protection with dexterity
Manufacturer: mapa™  541428
 View more versions of this product
Catalog No. 11-393-90B
  • / Pack of 100 Each
Description & Specifications
Specifications
Powdered Powdered
Color Yellow
Length (English) 10.5 in.
Thickness (English) 5 mil
Length (Metric) 26.7cm
Certifications CE
Exterior Finish Bisque
For Use With (Application) intricate assembly, film processing, biological research, Resin application, Epoxy layup and furniture finishing, liquid tight gloves outperform natural Rubber and Nitrile in Acids, Caustics, Alcohols, Aldehydes, Oils and G
Glove Cuff Style Pinked
Size Large
Hand Left
Material Vinyl