Please login to your online account to display your discounted pricing
MAPA™ Pylox™ Vinyl Bisque Grip Gloves
Combining chemical and mechanical protection with dexterity
Manufacturer: mapa™  541427
 View more versions of this product
Catalog No. 11-393-90A
  • / Case of 1000 Each
Description & Specifications
Specifications
Powdered Powdered
Length (Metric) 26.7cm
Length (English) 10.5 in.
Thickness (English) 5 mil
Color Yellow
Certifications CE
For Use With (Application) intricate assembly, film processing, biological research, Resin application, Epoxy layup and furniture finishing, liquid tight gloves outperform natural Rubber and Nitrile in Acids, Caustics, Alcohols, Aldehydes, Oils and G
Glove Cuff Style Pinked
Exterior Finish Bisque
Hand Left
Material Vinyl
Size Medium