Please login to your online account to display your discounted pricing

MAPA™ Two-Tone™ Flock-Lined Neoprene/Natural Rubber Gloves


Provide reinforced strength

Manufacturer: mapa™  405426

 View more versions of this product

Catalog No. 11-392-33A

  • / Dozen of 12 Pair
  • / Case of 6 Dozen
 

Description & Specifications

Specifications

Certifications 21 CFR 170-199
Glove Cuff Style Pinked
For Use With (Application) aircraft assembly, chemical processing, food processing, Fiber Glass lay-up, general maintenance
Exterior Finish Diamond
Thickness 20mil
Length (Metric) 33cm
Color Blue, yellow
Size 6
Material Neoprene, Natural Rubber Latex
Hand Hand-specific
Lining Cotton Flock
Length (English) 13 in.