Please login to your online account to display your discounted pricing

MAPA™ Two-Tone™ Flock-Lined Neoprene/Natural Rubber Gloves

Provide reinforced strength

$26.15 - $26.15

Specifications

Certifications 21 CFR 170-199
Color Blue
Color Yellow
Length (English) 13 in.
Length (Metric) 33cm
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

11-392-33A

 
mapa™
405426
6 Dozen of 12 Pair for $26.15
Case of 6 Dozen for $156.90

11-392-33B

 
mapa™
405427
7 Dozen of 12 Pair for $26.15
Case of 6 Dozen for $156.90

11-392-33C

 
mapa™
405428
8 Dozen of 12 Pair for $26.15
Case of 6 Dozen for $156.90

11-392-33D

 
mapa™
405429
9 Dozen of 12 Pair for $26.15
Case of 6 Dozen for $156.90

11-392-33E

 
mapa™
405420
10 Dozen of 12 Pair for $26.15
Case of 6 Dozen for $156.90
Description & Specifications

Specifications

Certifications 21 CFR 170-199
Color Blue
Color Yellow
Length (English) 13 in.
Length (Metric) 33cm
Material Latex, natural rubber
Material Neoprene
Exterior Finish Diamond
For Use With (Application) aircraft assembly, chemical processing, food processing, Fiber Glass lay-up, general maintenance
Glove Cuff Style Pinked
Hand Hand-specific
Lining Cotton Flock
Thickness (English) 20 mil

  • Double-dipped blue neoprene blend over yellow natural rubber latex
  • Recessed diamond grip on palm and fingers
  • Cotton flock lining absorbs perspiration and enhances user comfort; provides easy on-off
  • Two-color design provides a safety assurance wear indicator
  • Oil and chemical resistant
  • Length: 13 in. (33cm)
  • Thickness: 28mil

Comply with 21CFR170–199.