Please login to your online account to display your discounted pricing

MAPA™ Two-Tone™ Flock-Lined Neoprene/Natural Rubber Gloves


Provide reinforced strength

Manufacturer: mapa™  405420

 View more versions of this product

Catalog No. 11-392-33E

  • / Dozen of 12 Pair
  • / Case of 6 Dozen
 

Description & Specifications

Specifications

Certifications 21 CFR 170-199
Exterior Finish Diamond
Glove Cuff Style Pinked
For Use With (Application) aircraft assembly, chemical processing, food processing, Fiber Glass lay-up, general maintenance
Size 10
Thickness 20mil
Color Blue, yellow
Hand Hand-specific
Length (English) 13 in.
Length (Metric) 33cm
Lining Cotton Flock
Material Neoprene, Natural Rubber Latex