missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MAPA™ Temp-Tec™ Insulated Supported Neoprene Gloves
Protection from extreme temperatures
Manufacturer: MAPA™ 338608
Description
Protection from extreme temperatures
Ideal for applications involving hot sampling of petrochemicals and resins, food processing, hot steam cleaning and plastics molding and extruding.
- Constructed of nontoxic synthetic rubber
- Protect hands from discomfort during brief contact with hot metal to 480°F (249°C) and cold contact to -100°F (-73°C)
- Soft and flexible, with positive nonslip grip
- Integral 13 oz. cotton jersey thermal liner in palm area
- Two-color design provides a wear indicator to permit glove changeout before abrasion penetrates the shell
Compliance
FDA 510(k) approved; FDA 21 CFR 170-199 approved for food processing.
Specifications
Over edge | |
Insulated, Flocked | |
8 | |
17 in. | |
FDA 510(k), FDA 21 CFR 170-199 |
boiler room work, handling of hot piping, handling of moulds in the Ceramic industry, manufacture of adhesives, insulating electric wires, road works in cold conditions, steam cleaning, filleting fish, chemical treatment of Metals | |
Neoprene | |
Black,Blue | |
43.18 cm |