missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MAPA™ Technic/NS-450 Neoprene/Natural Rubber Gloves

Manufacturer:  MAPA™ 450441

 View more versions of this product

Catalog No. 19-050-575



Description

Description

Specifications

Specifications

30 mil
food processing, meat and poultry, chemical sampling, commercial housekeeping, inspection, Metal finishing, automotive painting, degreasing, plating, Bleaching, Leather tanning, commercial pot scrubbing, janitorial, maintenance
Neoprene,Rubber Latex
Black
16 in.
Flat
Cotton Flock
11
Non-sterile
40.64 cm
SDS
Documents