Please login to your online account to display your discounted pricing

MAPA™ Technic™ Flock-Lined Neoprene Gloves


Provides superior flexibility and tear strength

Manufacturer: mapa™  401449

 View more versions of this product

Catalog No. 11-392-34D

  • / Pair
  • / Dozen of 12 Pair
 

Description & Specifications

Specifications

Length (English) 12.2 in.
Length (Metric) 31cm
Size X-Large
Thickness 22mil (0.55mm)
Color Black