MAPA™ Technic™ Flock-Lined Neoprene Gloves
Provides superior flexibility and tear strength
Manufacturer:  MAPA™ 401440
 View more versions of this product
Catalog No. 11-392-34E
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
Embossed textured
food processing, meat and poultry, chemical sampling, commercial housekeeping, inspection, Metal finishing, automotive painting, degreasing, plating, Bleaching, Leather tanning, commercial pot scrubbing, janitorial, maintenance
Latex, natural rubber,Neoprene
22mil
Black
31.75cm
Contoured
Cotton Flock
10
USDA, 21 CFR 170-199, CE
12.5 in.