Please login to your online account to display your discounted pricing

MAPA™ StanZoil™ Knit-Lined Multi-Layer Supported Neoprene Gloves


Combine flexibility with abrasion, cut and tear resistance. MAPA™ StanZoil™ Knit-Lined Multi-Layer Supported Neoprene Gloves have unique multi-dip construction offers liquid-proof hand protection and excellent chemical resistance to most oils, grease, acids, caustics, alcohols, and solvents.

Manufacturer: mapa™  337428

 View more versions of this product

Catalog No. 11-388-118A

  • / Pair
  • / Dozen of 12 Pair
 

Description & Specifications

Specifications

Size 8
Certifications USDA, 21 CFR 170-199, CE
Glove Cuff Style Pinked
For Use With (Application) chemical processing, Petroleum refining, grinding and filing of castings, Metal finishing, general maintenance
Length (English) 14 in.
Length (Metric) 35.56cm
Lining Interlock Cotton knit
Material Neoprene
Coating Material Non-slip
Color Black, blue

  • Ideal for applications involving assembly, degreasing, lubricating, castings, steam cleaning, motor parts, chemical processing, petroleum refining, grinding, filing, metal finishing and general maintenance
  • Neoprene overdip in glove work area eliminates pinholes and weak spots throughout the glove and cuff
  • Nonslip finish ensures a safe grip—even on wet or oily parts
  • Two-way stretch cotton interlock knit lining insulates from heat and cold and absorbs perspiration
  • Seamless liner in work area protects hands from chafing
  • Roomy thumb crotch for long-wear comfort
  • Pinked cuff
  • Washable

Meet 21CFR 170-199.