Please login to your online account to display your discounted pricing

MAPA™ StanSolv™ Unlined Nitrile Gloves


Comfort and dexterity despite high mechanical strength

Manufacturer: mapa™  479410

 View more versions of this product

Catalog No. 11-392-2D

  • / Pair
  • / Dozen of 12 Pair
 

Description & Specifications

Specifications

Material Nitrile
Glove Cuff Style Flat
For Use With (Application) plating, chemical processing, meat and poultry processing, degreasing operations, Solvent washing, Acid etching
Powdered No
Coating Material Embossed
Thickness 15mil
Color Green
Length (Metric) 33cm
Size 10
Length (English) 13 in.