Please login to your online account to display your discounted pricing

MAPA™ StanSolv™ Unlined Nitrile Gloves


Comfort and dexterity despite high mechanical strength

Manufacturer: mapa™  479419

 View more versions of this product

Catalog No. 11-392-2C

  • / Pair
  • / Dozen of 12 Pair
 

Description & Specifications

Specifications

Size 9
Thickness (English) 15mil
Length (English) 13 in.
Powdered Powder-free
Length (Metric) 33cm
Color Green
For Use With (Application) plating, chemical processing, meat and poultry processing, degreasing operations, Solvent washing, Acid etching
Glove Cuff Style Flat
Material Nitrile
Coating Material Embossed