Please login to your online account to display your discounted pricing

MAPA™ StanSolv™ Unlined Nitrile Gloves

Comfort and dexterity despite high mechanical strength

$2.07 - $123.09

Specifications

Coating Material Embossed
Color Green
Material Nitrile
For Use With (Application) plating, chemical processing, meat and poultry processing, degreasing operations, Solvent washing, Acid etching
Glove Cuff Style Flat
View More Specs
Products
Catalog Number Mfr. No. Thickness Length (English) Length (Metric) Size Price Quantity    

11-392-1C

 
mapa™
470429
11mil 13 in. 33cm 9 Pair for $2.07
Dozen of 12 Pair for $23.76

11-392-1D

 
mapa™
470420
11mil 13 in. 33cm 10 Pair for $2.26
Dozen of 12 Pair for $25.92

11-392-1B

 
mapa™
470428
11mil 13 in. 33cm 8 Pair for $2.07
Dozen of 12 Pair for $23.76

11-392-1A

 
mapa™
470427
11mil 13 in. 33cm 7 Pair for $2.07
Dozen of 12 Pair for $23.76

11-392-1E

 
mapa™
470421
11mil 13 in. 33cm 11 Pair for $2.26
Dozen of 12 Pair for $25.92

11-392-2D

 
mapa™
479410
15mil 13 in. 33cm 10 Pair for $2.60
Dozen of 12 Pair for $29.88

11-392-2C

 
mapa™
479419
15mil 13 in. 33cm 9 Pair for $2.60
Dozen of 12 Pair for $29.88

11-392-2A

 
mapa™
479417
15mil 13 in. 33cm 7 Pair for $2.57
Dozen of 12 Pair for $29.52

11-392-2E

 
mapa™
479411
15mil 13 in. 33cm 11 Pair for $2.54
Dozen of 12 Pair for $29.16

11-392-4B

 
mapa™
474018
22mil 14 in. 35.5cm 8 Dozen of 12 Pair for $102.46
Case of 4 Dozen for $409.84

11-392-4A

 
mapa™
474017
22mil 14 in. 35.5cm 7 Dozen of 12 Pair for $75.91
Case of 4 Dozen for $303.64

11-392-4C

 
mapa™
474019
22mil 14 in. 35.5cm 9 Dozen of 12 Pair for $102.46
Case of 4 Dozen for $409.84

11-392-5A

 
mapa™
480439
22mil 18 in. 45.7cm 9 Pair for $10.37
Dozen of 12 Pair for $118.80

11-392-5C

 
mapa™
480421
22mil 18 in. 45.7cm 11 CALL N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

11-392-5B

 
mapa™
480420
22mil 18 in. 45.7cm 10 CALL CALL N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

11-392-2B

 
mapa™
479418
15mil 13 in. 33cm 8 Dozen of 12 Pair for $27.30
Case of 6 Dozen for $163.80
Description & Specifications

Specifications

Coating Material Embossed
Color Green
Material Nitrile
For Use With (Application) plating, chemical processing, meat and poultry processing, degreasing operations, Solvent washing, Acid etching
Glove Cuff Style Flat
Powdered No

  • Z-Patter grip channels liquids away from glove surface for an enhanced grip in wet applications
  • Nitrile compound provides resistance to snags, punctures, abrasions and cuts
  • Chlorinated finish provides easy on/off
  • Straight cuff