Please login to your online account to display your discounted pricing

MAPA™ StanSolv™ Unlined Nitrile Gloves


Comfort and dexterity despite high mechanical strength

Manufacturer: mapa™  474017

 View more versions of this product

Catalog No. 11-392-4A

  • / Dozen of 12 Pair
  • / Case of 4 Dozen
 

Description & Specifications

Specifications

Material Nitrile
Length (Metric) 35.5cm
For Use With (Application) plating, chemical processing, meat and poultry processing, degreasing operations, Solvent washing, Acid etching
Glove Cuff Style Flat
Thickness 22mil
Coating Material Embossed
Size 7
Powdered No
Color Green
Length (English) 14 in.