MAPA™ StanSolv™ Heavy-Weight Nitrile Gloves
Carboxylated nitrile compound resists cuts, punctures, and abrasions and contains no fillers.
$5.42 - $7.42
Specifications
Certifications 21 CFR 170-199, CE
Color Green,White
Length (English) 15.5 in.
Length (Metric) 39cm
Material Nitrile
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-015-125C MAPA™
493420
10 Pair for $7.42
Dozen of 12 Pair for $84.96
19-015-125A MAPA™
493428
8 Pair for $7.42
Dozen of 12 Pair for $84.96
19-015-125D MAPA™
493421
11 Pair for $5.42
Dozen of 12 Pair for $62.04
19-015-125B MAPA™
493429
9 Pair for $7.42
Dozen of 12 Pair for $84.96
Specifications
Description
Silicone-free for use in painting operations. Ideal for acid dipping, plating applications, metal finishing, solvent washing, chemical processing, oil production and refining, degreasing operations, and Haz Mat cleanup.
  • Double-dipped to prevent thin spots and pinholes common to single-dipped gloves
  • Safety green exterior, white interior layers provide visual indicator of wear-through
  • Embossed raised-diamond grip pattern prevents buildup of animal fats and other greasy solids
  • Available in 12 in. (30cm) and 15 1/2 in. (39cm) lengths
  • Flock-lined with flat cuff
Certifications
CE marked CE approved.
Compliance
Meet 21CFR170–199.
Description & Specifications
Specifications
21 CFR 170-199, CE
15.5 in.
Nitrile
Flat
22mil
Green,White
39cm
Silicone-free for use in painting operations, ideal for Acid dipping, plating applications, Metal finishing, Solvent washing, chemical processing, Oil production and refining, degreasing operations and hazmat cleanup
Flock