Please login to your online account to display your discounted pricing

MAPA™ PROtector™ Nitrile-Natural Rubber Gloves


Green natural rubber with a blue nitrile blend overdip in the work area

Manufacturer: mapa™  293428

 View more versions of this product

Catalog No. 11-388-115C

  • / Pair
  • / Dozen of 12 Pair
 

Description & Specifications

Specifications

Material Nitrile
Material Rubber, natural
Thickness (English) 28mil
Length (Metric) 33cm
Color Blue
Color Green
Length (English) 13 in.
Liner Material Cotton Flock
Glove Cuff Style Straight
For Use With (Application) food handling and pharmaceutical applications
Size Large
Coating Material Embossed chained pattern