missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MAPA™ NiProTect™ Controlled Environment Nitrile Gloves

Critically Clean 100% nitrile formulation.

$24.76 - $25.61

Specifications

Classification ISO Class 4
Color White
Length (English) 12 in.
Length (Metric) 30.48cm
Material Nitrile
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-050-338D MAPA™
529428
Large Pack of 100 Each for $25.61
Case of 10 Pack for $248.00
19-050-338C MAPA™
529427
Medium Pack of 100 Each for $24.76
Case of 10 Pack for $239.80
Specifications

Description

Meet the individual cleanliness level of your controlled environment. The 100% nitrile formulation eliminates protein allergy concerns while exhibiting excellent ESD properties and chemical splash protection. At 4mil thick, this glove delivers maximum tactility with good durability. Textured fingertips and unique finish enhance wet and dry gripping. Rolled cuff edge reduces glove roll-down and tearing when donning.
Available in critically clean (CC) for ISO Class 4 (formerly Class 10) compatibility. Ambidextrous.

Order Info

Sold in packs of 100.

Compliance

Meet or exceed ASTM D 3578 and ASTM D 130.

Description & Specifications

Specifications

ISO Class 4
12 in.
Nitrile
Rolled
4 mil
White
30.48cm
Textured
Ambidextrous