Please login to your online account to display your discounted pricing

MAPA™ Fluonit Gloves

Tactile-sensitivity gloves with wear indicator

$72.39 - $72.39

Specifications

Color Black
Length (English) 12 in.
Length (Metric) 30.5cm
Material Fluoroelastomer
Exterior Finish Smooth
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-165-705

 
mapa™
468128
8 Pair for $72.39
Description & Specifications

Specifications

Color Black
Length (English) 12 in.
Length (Metric) 30.5cm
Material Fluoroelastomer
Exterior Finish Smooth
For Use With (Application) Acids, Alkali compounds, Alcohols, High Aromatic Solvents, Aliphatics and Chlorines
Glove Cuff Style Pinked
Thickness (English) 20 mil

  • Excellent flexibility and thickness optimization due to Mapa multi-layer technology
  • Good dexterity and comfort
  • Nitrile inner layer for excellent mechanical resistance and a longer service life
  • Green inner layer contains wear indicator for greater user safety
  • Length: 30.5cm (12 in.)
  • Color: Black

Site pollution control; printing; petrochemical refining; metal degreasing and etching