missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MAPA™ Blue-Grip™ LL-301 Heavyweight Latex Gloves, Interlock Lining
Heavy gauge, 45 mil thick rubber latex gloves provide mechanical and chemical protection, high cut resistance and tear strength.
Supplier: MAPA™ 301426
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Description
Rated excellent for handling acids, “mild” caustics, alcohols, sanitizing agents and detergents, as well as other chemical, mechanical and thermal hazards. Ideal for handling frozen food, hot containers, sharp edges, abrasives, cutting and tearing hazards, sheet metal and plastic resins.
- Heavy-duty, liquid-proof gloves with a double-dipped nonabrasive, nonslip grip
- Interlock-knit cotton lining absorbs perspiration
- For intermittent exposure from -17° to +100°C (0° to 212°F)
- Gloves feature a natural rubber latex coating firmly bonded to a reinforcing, 9 oz. (255g) cotton knit lining
- Hand contoured with no seams in the work area
- Light blue with a medium blue grip
- Launderable
Specifications
21 CFR 170-199 | |
No | |
Small | |
1.14 mm | |
Pinked | |
Interlock knit | |
301426 | |
30.5 cm |
Blue | |
Latex | |
45 mil | |
Handling acids, “mild” caustics, alcohols, sanitizing agents and detergents, as well as other chemical, mechanical and thermal hazards, handling frozen food, hot containers, sharp edges, abrasives, cutting and tearing hazards, sheet metal and Pl | |
Etched | |
Lined | |
12 in. |