Please login to your online account to display your discounted pricing
MAPA™ Blue-Grip™ Latex Gloves
Provides easy on-off thermal protection
Manufacturer: mapa™  301428
 View more versions of this product
Catalog No. 11-392-40C
  • / Pair
  • / Case of 48 Pair
Description & Specifications
Specifications
Material Rubber, natural
Thickness (English) 13mil
Color Blue
Length (English) 12 in.
Length (Metric) 30.5cm
Certifications 21 CFR 170-199
Glove Cuff Style Pinked
For Use With (Application) handling Acids, “mild” Caustics, Alcohols, sanitizing agents and Detergents, as well as other chemical, mechanical and thermal hazards, handling frozen food, hot containers, sharp edges, abrasives, cutting and tearing hazards, sheet Metal and Pl
Size Large
Exterior Finish Non-slip
Lining Interlock knit