missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MAPA™ Optinit Nitrile Gloves

$1.20 - $11.95

Specifications

Color Blue
Length (English) 12 in.
Length (Metric) 30.5cm
Material Nitrile
For Use With (Application) laboratory and food application where dexterity, plus mild chemical/liquid protection is required
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-130-2641A MAPA™
472416
6 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-130-2641B MAPA™
472417
7 Pair for $1.20
Pack of 10 Pair for $12.00
19-130-2641C MAPA™
472418
8 Pair for $1.20
Pack of 10 Pair for $12.00
19-130-2641D MAPA™
472419
9 Pair for $1.20
Pack of 10 Pair for $12.00
Specifications Description & Specifications

Specifications

Blue
30.5cm
laboratory and food application where dexterity, plus mild chemical/liquid protection is required
Powder-free
12 in.
Nitrile
Flat
8mil