missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MAPA™ Krytech 576 Cut-Resistant Gloves, Palm-Coated

High-performance cut protection gloves offer comfort and dexterity for intricate handling in dry or oily environments.

$8.20 - $8.20

Specifications

Certifications/Compliance (Europe) Cat. 2 CE, EN 420
Construction Seamless Knit
Color Gray
Cut Resistance Rating ANSI A4
Hand Specific
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19249780
View Documents Promotion Details
MAPA™
576417
7
Pair for $8.20
Case of 96 Pair for $697.92
 
Description

Description

Ideal for automotive and general manufacturing, handling metal parts, assembly of thin metal sheets, finishing sharp parts, plastic industry and post-injection material removal

  • Polyurethane coating on palm and fingers
  • Smooth, knit wrist
  • Color: Gray

Specifications

Specifications

Cat. 2 CE, EN 420
Gray
Specific
Cut Resistant Gloves
Handling Metal parts, assembly of thin Metal sheets, finishing sharp parts
KryTech 576
27.9 cm
Seamless Knit
ANSI A4
HDPE
EN 388:4X42D
Knit
11 in.
SDS
missing translation for 'documents'

missing translation for 'documents'