Please login to your online account to display your discounted pricing

MAPA™ Krynit™ 563 Cut-Resistant Gloves

Provides cut/liquid protection for intricate handling in oily environments

$14.11 - $14.11

Specifications

Color Black
Color Gray
Cut Resistance Rating Level 2
Material HDPE (High-density Polyethylene)
Material Polyamide
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity    

19-249-772

 
mapa™
563418
9.35 in. 23.7cm 8 Pair for $14.11
Case of 96 Pair for $1,242.24

19-249-773

 
mapa™
563419
9.85 in. 25cm 9 Pair for $14.11
Case of 96 Pair for $1,242.24

19-249-771

 
mapa™
563417
9 in. 22.8cm 7 Pair for $14.11
Case of 96 Pair for $1,242.24

19-249-774

 
mapa™
563410
9.85 in. 25cm 10 Pair for $14.11
Case of 96 Pair for $1,242.24

19-249-775

 
mapa™
563411
10.5 in. 26cm 11 Pair for $14.11
Case of 96 Pair for $1,242.24
Description & Specifications

Specifications

Color Black
Color Gray
Cut Resistance Rating Level 2
Material HDPE (High-density Polyethylene)
Material Polyamide
For Use With (Application) mechanical maintenance, stacking Oily panels, pressing Metal, handling Metal sheets
Glove Cuff Style Knit
Hand Hand-specific

  • Nitrile coated palm and finger
  • Smooth grip
  • Color coded knit wrist
  • HDPE and polyamide 13-gauge liner with nitrile coating provides abrasion resistance along with comfort
  • Color: Gray with black coating
  • Launderable
  • Ideal for places where exposure could cause slight cuts like handling small sharp objects
  • Length: 9 to 10.5 in.

  • Automotive/ mechanical industries
  • Mechanical maintenance
  • Handling oily panels
  • Pressing metal, handling metal sheets
  • Paper industry
  • Handling cardboard, paper
  • Utility knife handling

ANSI cut level 2 and EN cut level 3