Please login to your online account to display your discounted pricing
MAPA™ Classics™ L-200 Gloves
Prevents greasy solids build-up
$1.09 - $16.02
Products
Catalog Number Mfr. No. Price Quantity    
19-042-650
mapa™
158569
CALL CALL N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
19-999-972D
mapa™
177018
Dozen for $16.02
Case of 12 Dozen for $183.72
Description & Specifications
Resistant to mild acids, alcohols, alkalis, salts, aldehydes and some animal and vegetable fats. Excellent resistance to abrasions, cuts and tears. For use in food applications, electronic assembly, lab and agricultural chemical handling, glassware handling and commercial housekeeping.
  • Unsupported natural rubber gloves are unlined with a chlorinated powder-free finish
  • Raised diamond-grip pattern prevents greasy solids buildup and provides a positive grip on hard-to-grasp materials
  • Shaped to reduce fatigue with contoured palm design and curved fingers
  • With flat cuff
  • Length: 12 in. (30cm); thickness: 18mil
  • Color, dark blue
CE marked CE approved.
Meet 21CFR170-199,