Please login to your online account to display your discounted pricing

MAPA™ AdvanTech™ StanSolv™ Nitrile Gloves


Manufacturer-suggested ISO Class 4

Manufacturer: mapa™  519318

 View more versions of this product

Catalog No. 11-392-7B

  • / Pair
  • / Dozen of 12 Pair
 

Description & Specifications

Specifications

Classification ISO Class 4
Length (English) 13 in.
Length (Metric) 33.02cm
Material Nitrile
Thickness 11mil
Size 8
Color Green