missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MAPA™ AdvanTech™ Rollpruf™ Special Formula Gloves

Exclusive cuff design prevents rolldown

Manufacturer:  MAPA™ 513984

 View more versions of this product

Catalog No. 11-394-69


 

Specifications Description & Specifications

Specifications

Yes
Rolled
No
7.5
Powdered
12 in.
Textured
semiconductor, electronics, aerospace, pharmaceutical and biotechnology industries, no pigments, fillers or DOP Plasticizers, resist Acids, Salts, Alcohols, Ketones and Alkalis
Tri-polymer
8mil
Black
30.5cm