Please login to your online account to display your discounted pricing
MAPA™ AdvanTech™ Rollpruf™ Special Formula Gloves
Exclusive cuff design prevents rolldown
Manufacturer: mapa™  513562
 View more versions of this product
Catalog No. 11-394-86
  • / Case of 200 Pair
Description & Specifications
Specifications
Color Blue
Length (English) 12 in.
Powdered Powdered
Thickness (English) 8mil
Length (Metric) 30.5cm
Glove Cuff Style Rolled
For Use With (Application) semiconductor, electronics, aerospace, pharmaceutical and biotechnology industries, no pigments, fillers or DOP Plasticizers, resist Acids, Salts, Alcohols, Ketones and Alkalis
Exterior Finish Textured
Latex Free No
Size 9.5
Material Tri-polymer
Disposable Yes