Invitrogen™ MAP4K5 (KHS1) Recombinant Human Protein DFS Item
MAP4K5 (KHS1) is a serine/threonine kinase that belongs to the GCK family of kinases.
Manufacturer:  Invitrogen™ LSPV4014
 View more versions of this product
Catalog No. PV4014
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
MAP4K5 (KHS1) has been implicated as an upstream regulator of MAP kinase signaling pathways.
Specifications
Specifications
NP_942089
Kinase Assay
Frozen
122.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
STE20 Kinase Family
Yes
Baculovirus
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MAP4K5 (KHS1)
Human
GST Tag
Documents