missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ MAP4K5 (KHS1) Recombinant Human Protein DFS Item

MAP4K5 (KHS1) is a serine/threonine kinase that belongs to the GCK family of kinases.

$3,050.00 - $3,050.00

Specifications

Format Frozen
Host Species Human
Protein Tag GST Tag
Recombinant Yes
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR6671A View Documents Invitrogen™
LSPR6671A
100μg Each for $3,050.00
 
PV4014 View Documents Invitrogen™
LSPV4014
1mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

MAP4K5 (KHS1) has been implicated as an upstream regulator of MAP kinase signaling pathways.

Specifications

Specifications

Frozen
Human
GST Tag
Yes