Invitrogen™ MAP4K4 (HGK) Recombinant Human Protein DFS Item
MAP4K4 (HGK) is a serine/threonine kinase.
Manufacturer:  Invitrogen™ LSPV4013
 View more versions of this product
Catalog No. PV4013
Description
Description
MAP4K4 (HGK) plays an important role in stress-activated MAPK core signaling pathways.
Specifications
Specifications
NP_004825
Frozen
65kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
STE20 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MAP4K4 (HGK)
Baculovirus
Human
GST Tag
Documents