Please login to your online account to display your discounted pricing

MAP4, Tocris

Potent group II mGlu agonist. Also specific group III mGlu antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material MAP4
Assay Percent Range >96%
Molecular Formula C5H12NO5P
Formula Weight 197.13
Synonym (S)-2-Amino-2-methyl-4-phosphonobutanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

071110

 
r & d systems
RDS071110
10mg Each for $252.39

071150

 
r & d systems
RDS071150
50mg Each for $1,050.42
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 197.13
Chemical Name or Material MAP4
Molecular Formula C5H12NO5P
Synonym (S)-2-Amino-2-methyl-4-phosphonobutanoic acid
CAS 157381-42-5
Assay Percent Range >96%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature