Invitrogen™ MAP3K9 (MLK1) Recombinant Human Protein DFS Item
MAP3K9 (MLK1) is a serine/threonine protein kinase.
Manufacturer:  Invitrogen™ LSPV4012
 View more versions of this product
Catalog No. PV4012
Description
Description
MAP3K9 (MLK1) functions in the MAP kinase pathway.
Specifications
Specifications
NP_149132
Frozen
83.4kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
MLK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MAP3K9 (MLK1)
Baculovirus
Human
GST Tag
Documents