Invitrogen™ MAP3K8 (COT) Recombinant Human Protein DFS Item
MAP3K8 (COT) is a serine/threonine kinase.
Manufacturer:  Invitrogen™ LSPV4330
 View more versions of this product
Catalog No. PV4330
Description
Description
MAP3K8 can activate both the MAP and JNK kinase pathways and plays a role in the cell cycle.
Specifications
Specifications
NP_005195
Kinase Assay
Frozen
69.9kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
STE Unique Kinase Family
Yes
Baculovirus
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
1mg
RUO - research use only
MAP3K8 (COT)
Human
GST Tag