missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ MAP3K8 (COT) Recombinant Human Protein DFS Item

MAP3K8 (COT) is a serine/threonine kinase.

$1,976.00 - $8,390.00

Specifications

Format Frozen
Host Species Human
Protein Tag GST Tag
Recombinant Yes
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR7887A View Documents Invitrogen™
LSPR7887A
100μg Each for $1,976.00
 
PV4330 View Documents Invitrogen™
LSPV4330
1mg Each for $8,390.00
 
Description

Description

MAP3K8 can activate both the MAP and JNK kinase pathways and plays a role in the cell cycle.

Specifications

Specifications

Frozen
Human
GST Tag
Yes