Invitrogen™ MAP3K5 (ASK1) Recombinant Human Protein DFS Item
MAP3K5 (ASK1) is a serine/threonine kinase.
Manufacturer:  Invitrogen™ LSPV4011
 View more versions of this product
Catalog No. PV4011
Description
Description
MAP3K5 (ASK1) is abundantly expressed in human heart and pancreas.
Specifications
Specifications
NP_005914
Frozen
182kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
STE11 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MAP3K5 (ASK1), ASK1
Baculovirus
Human
GST Tag
Documents