MAP3K5 (ASK1) Recombinant Human Protein DFS Item
MAP3K5 (ASK1) Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3809
Catalog No. PV3809
Description
Description
MAP3K5 (ASK1) is a serine⁄threonine kinase that is abundantly expressed in human heart and pancreas.
Specifications
Specifications
NP_005914
Cell viability, proliferation and function, pharma and biopharma, protein assays and analysis, target and lead identification and validation
182kDa
80% by SDS-PAGE
–80°C
1 Each
MAP3K5 (ASK1), ASK1
MAP3K5 (ASK1) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 8.0), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 333μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
STE11
60nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL
GST Tag
0.2mg/mL
4217, 4776
MAP3K5 (ASK1) Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Serine/threonine kinase
MAP3K5 (ASK1) Recombinant Human Protein
Kinase assay
Baculovirus
MAP3K5 (ASK1) Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.