Invitrogen™ MAP3K3 (MEKK3) Recombinant Human Protein DFS Item
MAP3K3 (MEKK3) phosphorylates multiple MAP2Ks.
Manufacturer:  Invitrogen™ LSPV4194
 View more versions of this product
Catalog No. PV4194
Description
Description
MAP3K3 (MEKK3) directly regulates MAPK8 and extracellular signal-regulated protein kinase (ERK) pathways.
Specifications
Specifications
NP_002392
Frozen
99.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
STE11 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MAP3K3 (MEKK3)
Baculovirus
Human
GST Tag
Documents