missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Human MAP3K14 (NIK), GST Tag Recombinant Protein
Human MAP3K14 (NIK), GST Tag Recombinant Protein
$770.00 - $11270.00
Specifications
Accession Number | NP_003945.2 |
---|---|
Gene Alias | MAP3K14 (NIK) |
KinaseFamily | STE Unique Kinase Family |
Species | Human |
Recombinant | Recombinant |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR8920A
|
Invitrogen™
PR8920A |
100μg |
Each for $2,650.00
|
|
|||||
PV4903
|
Invitrogen™
PV4903 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
PV4902
|
Invitrogen™
PV4902 |
10 μg |
Each for $770.00
|
|
|||||
Description
MAP3K14 phosphorylates and activates CHUK (IKKalpha) and IKBKB (IKK beta), which in turn phosphorylate IkB-alpha, releasing NF-KappaB leading to up-regulation of inflammatory response genes.
Specifications
NP_003945.2 | |
STE Unique Kinase Family | |
Recombinant | |
Baculovirus |
MAP3K14 (NIK) | |
Human | |
GST Tag |