Invitrogen™ MAP3K14 (NIK) Recombinant Human Protein DFS Item
MAP3K14 (NIK) is structurally related to MAP3Ks.
Manufacturer:  Invitrogen™ LSPV4903
 View more versions of this product
Catalog No. PV4903
Description
Description
MAP3K14 phosphorylates and activates CHUK (IKKalpha) and IKBKB (IKK beta), which in turn phosphorylate IkB-alpha, releasing NF-KappaB leading to up-regulation of inflammatory response genes.
Specifications
Specifications
NP_003945.2
Frozen
96.9kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
STE Unique Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol
1mg
RUO - research use only
MAP3K14 (NIK)
Baculovirus
Human
GST Tag
Documents