Invitrogen™ MAP3K11 (MLK3) Recombinant Human Protein DFS Item
MAP3K11 (MLK3) is a serine/threonine kinase.
Manufacturer:  Invitrogen™ LSPV4009
 View more versions of this product
Catalog No. PV4009
Description
Description
MAP3K11 (MLK3) activates both the p38 MAP kinase and c-Jun kinase pathways.
Specifications
Specifications
NP_002410.1
Frozen
120.1kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
MLK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MAP3K11 (MLK3)
Baculovirus
Human
GST Tag
Documents