Invitrogen™ MAP3K10 (MLK2) Recombinant Human Protein DFS Item
MAP3K10 (MLK2) is a serine/threonine kinase which has been shown to activate MAPK8/JNK and MKK4/SEK1.
Manufacturer:  Invitrogen™ LSPV4195
 View more versions of this product
Catalog No. PV4195
Description
Description
The MAP3K10 (MLK2) kinase itself can be phoshorylated, and is thus activated by JNK kinases. MAP3K10 functions preferentially on the JNK signaling pathway, and is reported to be involved in nerve growth factor (NGF) induced neuronal apoptosis.
Specifications
Specifications
NP_002437
Frozen
131.2kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
MLK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MAP3K10 (MLK2)
Baculovirus
Human
GST Tag
Documents