Invitrogen™ MAP2K3 (MEK3) Recombinant Human Protein DFS Item
MAP2K3 (MEK3) is a dual-specificity protein kinase which acts in cellular signal transduction pathways.
Manufacturer:  Invitrogen™ LSPV4006
 View more versions of this product
Catalog No. PV4006
Description
Description
MAP2K3 (MEK3) is necessary for the expression of glucose transporter.
Specifications
Specifications
NP_002747
Frozen
64.2kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
STE7 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MAP2K3 (MEK3)
Baculovirus
Human
GST Tag
Documents