missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MAP2 Mouse, eFluor 570, Clone: AP20, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS41976380

 View more versions of this product

Catalog No. 501130704



Description

Description

The monoclonal antibody AP20 recognizes the high molecular weight isoforms of microtubule-associated protein-2 (MAP2a and MAP2b) of human, rat, mouse, bovine, chicken, and xenopus. MAP2 belongs to a family of heat-stable phosphoproteins that are differentially expressed during brain development. MAP2 expression is localized to the cell body and dendrites of neurons and is involved in stabilizing microtubules by crosslinking with intermediate filaments. Expression of MAP2 is commonly used as a neural marker given that its expression becomes more pronounced with neuronal maturation.
Specifications

Specifications

MAP2
AP20
eFluor 570
Conjugated
Murine
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Microscopy
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Microtubule-associated protein 2
IgG1, kappa
RUO
Primary
Bovine, Chicken, Human, Murine, Rat, Xenopus
Documents

For Research Use Only.