Please login to your online account to display your discounted pricing

MAP2 Mouse, Clone: AP20, eBioscience™

Mouse Monoclonal Antibody

$117.45 - $294.35

Specifications

Antigen MAP2
Clone AP20
Host Species Murine
Gene Alias Microtubule-associated protein 2
Species Reactivity Bovine, Chicken, Human, Mouse, Rat, Xenopus
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122839

 
life technologies
LS14976380
25μg Each for $117.45

501122840

 
life technologies
LS14976382
100μg Each for $294.35
Description & Specifications

Specifications

Antigen MAP2
Clone AP20
Host Species Murine
Gene Alias Microtubule-associated protein 2
Species Reactivity Bovine, Chicken, Human, Mouse, Rat, Xenopus
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Applications Microscopy
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody AP20 recognizes the high molecular weight isoforms of microtubule-associated protein-2 (MAP2a and MAP2b) of human, rat, mouse, bovine, chicken, and xenopus. MAP2 belongs to a family of heat-stable phosphoproteins that are differentially expressed during brain development. MAP2 expression is localized to the cell body and dendrites of neurons and is involved in stabilizing microtubules by crosslinking with intermediate filaments. Expression of MAP2 is commonly used as a neural marker given that its expression becomes more pronounced with neuronal maturation.